Merck
  • N-carbamylglutamate enhances ammonia detoxification in a patient with decompensated methylmalonic aciduria.
N-carbamylglutamate enhances ammonia detoxification in a patient with decompensated methylmalonic aciduria.
Molecular genetics and metabolism (2003-09-02)
Boris Gebhardt, Stefan Vlaho, Doris Fischer, Adrian Sewell, Hansjosef Böhles
ABSTRACT
In patients with methylmalonic aciduria (MMA), the accumulating metabolite propiony-CoA results in an inhibition of the urea circle via the decreased synthesis of N-acetylglutamate, an essential activator of carbamylphosphat synthetase (CPS). This results in one of the major clinical problems which is hyperammonaemia. In a patient with decompensated MMA, the CPS activator carbamylglutamate was tested for its ability to antagonize the propionyl-CoA-induced hyperammonaemia. Oral carbamylgutamate administration resulted in an impressive increase in ammonia detoxification compared to peritoneal dialysis. Safe, fast and easy to administer, carbamylglutamate improves the acute therapy of decompensated MMA by increasing ammonia detoxification and avoiding hyperammonaemia.
Sigma-Aldrich
N-Carbamyl-L-glutamic acid
Pricing and availability is not currently available.