Merck
  • In situ UV-visible spectroelectrochemical studies on the copolymerization of diphenylamine with ortho-methoxy aniline.
In situ UV-visible spectroelectrochemical studies on the copolymerization of diphenylamine with ortho-methoxy aniline.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-04-26)
P Santhosh, A Gopalan, T Vasudevan
ABSTRACT
UV-visible spectroelectrochemical studies on copolymerization of diphenylamine (DPA) with ortho-methoxy aniline (OMA) were carried out for different feed ratios of DPA and OMA using indium tin oxide (ITO)-coated glass as working electrode. The UV-visible spectra show clear dependencies on the molar feed composition of DPA or OMA used in electropolymerization. Derivative cyclic voltabsorptogram (DCVA) was deduced at the wavelengths corresponding to the absorption by the intermediate species and used to confirm the intermediates generated during the electropolymerization. The composition of DPA and OMA in the copolymer for the copolymers synthesized with different molar feed ratios of DPA and OMA was determined by UV-visible spectroscopy. Reactivity ratios of DPA and OMA were deduced by using Fineman-Ross and Kelen-Tudos methods and correlated with spectroelectrochemical results.
Sigma-Aldrich
o-Anisidine, ≥99%
Pricing and availability is not currently available.