Merck
  • Ph2SO/Tf2O: a powerful promotor system in chemoselective glycosylations using thioglycosides.
Ph2SO/Tf2O: a powerful promotor system in chemoselective glycosylations using thioglycosides.
Organic letters (2003-04-26)
Jeroen D C Codée, Remy E J N Litjens, René den Heeten, Herman S Overkleeft, Jacques H van Boom, Gijs A van der Marel
ABSTRACT
Diphenylsulfoxide in combination with triflic anhydride provides a very potent thiophilic glycosylation promotor system, capable of activating disarmed thioglycosides. The usefulness of this novel thiophilic activator is illustrated in a successful chemoselective glycosylation sequence in which the donor thioglycoside in the first condensation step may be either armed or disarmed. [reaction: see text]
Sigma-Aldrich
Trifluoromethanesulfonic anhydride, purum, ≥98.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Diphenyl sulfoxide, 96%
Pricing and availability is not currently available.