Merck
  • "Untypical aging off-flavor" in wine: synthesis of potential degradation compounds of indole-3-acetic acid and kynurenine and their evaluation as precursors of 2-aminoacetophenone.
"Untypical aging off-flavor" in wine: synthesis of potential degradation compounds of indole-3-acetic acid and kynurenine and their evaluation as precursors of 2-aminoacetophenone.
Journal of agricultural and food chemistry (2002-07-11)
Katrin Hoenicke, Ole Borchert, Kai Grüning, Thomas J Simat
ABSTRACT
Kynurenine (1) and indole-3-acetic acid (2) are considered as potential precursors of 2-aminoacetophenone (3), which is regarded to be the aroma impact compound causing an "untypical aging off-flavor" (UTA) in Vitis vinifera wines. The mechanism of the formation of 3 was studied using model fermentation and model sulfuration media spiked with 1 or 2 as potential precursors. Possible degradation products such as kynurenamine (4) and kynurenic acid (5), or skatole (6), 2-oxoskatole (7), 2-formamidoacetophenone (8), 2-oxindole-3-acetic acid (9), and 3-(2-formylaminophenyl)-3-oxopropionic acid (10) were evaluated by HPLC-UV of the fermentation and sulfuration media and comparison with synthesized 7, 8, 9, and 10. The synthesis of the possible precursor 4-(2-aminophenyl)-2,4-dioxobutanoic acid (11), a proposed metabolite of 1 failed because a spontaneous cyclization yields 5 and N-oxo-kynurenic acid (12), but not 11. It could be shown that the formation of 3 is triggered by an oxidative degradation of 2 after sulfuration with potassium bisulfite via the intermediates 10 and 8. However, no formation of 3 occurred during sulfuration of a model wine spiked with 1 or during fermentation of a model must spiked with 1 or 2.
Sigma-Aldrich
2′-Aminoacetophenone, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
2′-Aminoacetophenone, ≥98%
Pricing and availability is not currently available.