Merck
  • An oligomer from flaxseed composed of secoisolariciresinoldiglucoside and 3-hydroxy-3-methyl glutaric acid residues.
An oligomer from flaxseed composed of secoisolariciresinoldiglucoside and 3-hydroxy-3-methyl glutaric acid residues.
Phytochemistry (2001-09-29)
A Kamal-Eldin, N Peerlkamp, P Johnsson, R Andersson, R E Andersson, L N Lundgren, P Aman
ABSTRACT
A straight-chain oligomeric structure composed of five secoisolariciresinoldiglucoside (SDG) residues interconnected by four 3-hydroxy-3-methyl glutaric acid (HMGA) residues (molecular weight ca. 4000 Da) was assigned to the main lignan of flaxseed on the basis of nuclear magnetic resonance spectroscopy (NMR).
Sigma-Aldrich
3-Hydroxy-3-methylglutaric acid, ≥95%