Merck
Synthesis and antiperoxidant activity of new phenolic O-glycosides.
Carbohydrate research (2001-03-28)
F Ponticelli, A Trendafilova, M Valoti, S Saponara, G Sgaragli
ABSTRACT
We describe the synthesis of some 3-tert-butyl-4-hydroxyphenyl D-glycopyranosides by reaction of tert-butylhydroquinone with beta-D-pentaacetyl-glucose, beta-D-pentaacetyl-galactose, 2-acetamido- and 3,4,6-tri-O-acetyl-2-butanamido-2-deoxy-beta-D-glucopyranosyl chlorides as well as the formation of anomeric 3-tert-butyl-4-hydroxyphenyl 4,6-di-O-acetyl-2,3-dideoxy-D-erythro-hex-2-eno-pyranosides by reaction between tert-butylhydroquinone and 3,4,6-tri-O-acetyl-D-glucal. All compounds, except 3-tert-butyl-4-hydroxyphenyl alpha- and beta-D-glucopyranosides, inhibited lipid peroxidation with a degree of potency comparable to that of tert-butyl hydroxyanisole.
Sigma-Aldrich
β-D-Glucose pentaacetate, 98%
Sigma-Aldrich
α-D(+)-Glucose pentaacetate, 99%