Merck
  • Indirect atomic absorption spectrometric determination of pindolol, propranolol and levamisole hydrochlorides based on formation of ion-associates with ammonium reineckate and sodium cobaltinitrite.
Indirect atomic absorption spectrometric determination of pindolol, propranolol and levamisole hydrochlorides based on formation of ion-associates with ammonium reineckate and sodium cobaltinitrite.
Journal of pharmaceutical and biomedical analysis (2000-03-17)
S Khalil, N Borham
ABSTRACT
A new simple, accurate, precise and sensitive indirect method for the determination of pindolol HCl (1), propranolol HCl (2) and levamisole HCl (3) using atomic absorption spectrometry has been developed. The method is based on precipitation of the ion-associates formed from the reaction of (1), (2) or (3) with ammonium reineckate and/or sodium cobaltinitrite. The solubility of the solid complexes at the optimum conditions of pH and ionic strength values have been studied. Saturated solutions of each ion-associate were prepared under the optimum conditions and the metal ion content in the supernatant was determined. The method has been used for the determination of 1.14-17.07, 1.18-17.75 and 1.08-16.24 microg/ml of (1), (2) and (3), respectively, using ammonium reineckate, and 1.71-25.60, 1.77-26.62 and 1.62-24.36 microg/ml of (1), (2) and (3), respectively, using sodium cobaltinitrite. The method developed was applied for analysis of bulk drugs and some of their pharmaceutical preparations.
Sigma-Aldrich
Reinecke salt, ACS reagent, ≥93.0%
Sigma-Aldrich
Sodium hexanitrocobaltate(III), ACS reagent
Supelco
Levamisole Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material