Merck
  • Purification and molecular cloning of the APO-1 cell surface antigen, a member of the tumor necrosis factor/nerve growth factor receptor superfamily. Sequence identity with the Fas antigen.
Purification and molecular cloning of the APO-1 cell surface antigen, a member of the tumor necrosis factor/nerve growth factor receptor superfamily. Sequence identity with the Fas antigen.
The Journal of biological chemistry (1992-05-25)
A Oehm, I Behrmann, W Falk, M Pawlita, G Maier, C Klas, M Li-Weber, S Richards, J Dhein, B C Trauth
ABSTRACT
The APO-1 antigen as defined by the mouse monoclonal antibody anti-APO-1 was previously found to be expressed on the cell surface of activated human T and B lymphocytes and a variety of malignant human lymphoid cell lines. Cross-linking of the APO-1 antigen by anti-APO-1 induced programmed cell death, apoptosis, of APO-1 positive cells. To characterize the APO-1 cell surface molecule and to better understand its role in induction of apoptosis, the APO-1 protein was purified to homogeneity from membranes of SKW6.4 B lymphoblastoid cells by solubilization with sodium deoxycholate, affinity chromatography with anti-APO-1 antibody, and reversed phase high performance liquid chromatography. Each purification step was followed by an APO-1-specific solid phase enzyme-linked immunosorbent assay using the monoclonal antibody anti-APO-1. In sodium dodecyl sulfate-polyacrylamide gel electrophoresis, the APO-1 antigen was found to be a membrane glycoprotein of 48-kDa. Endoproteinase-cleaved peptides of the APO-1 protein were subjected to amino acid sequencing, and corresponding oligonucleotides were used to identify a full-length APO-1 cDNA clone from an SKW6.4 cDNA library. The deduced amino acid sequence of APO-1 showed sequence identity with the Fas antigen, a cysteine-rich transmembrane protein of 335 amino acids with significant similarity to the members of the tumor necrosis factor/nerve growth factor receptor superfamily. The APO-1 antigen was expressed upon transfection of APO-1 cDNA into BL60-P7 Burkitt's lymphoma cells and conferred sensitivity towards anti-APO-1-induced apoptosis to the transfectants.
Sigma-Aldrich
Fas/Fc Chimera from mouse, >97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture