Merck
  • GC-MS investigation and acetylcholinesterase inhibitory activity of Galanthus rizehensis.
GC-MS investigation and acetylcholinesterase inhibitory activity of Galanthus rizehensis.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2013-07-04)
Buket Bozkurt Sarikaya, Nehir Unver Somer, Gulen Irem Kaya, Mustafa Ali Onur, Jaume Bastida, Strahil Berkov
ABSTRACT
GC-MS (gas chromatography-mass spectrometry) analyses of alkaloids in the aerial parts and bulbs of Galanthus rizehensis Stern (Amaryllidaceae), collected during two different vegetation periods, was performed. Twenty three alkaloids were identified in four different alkaloid extracts. Acetylcholinesterase (AChE) inhibitory activities of the alkaloid extracts were tested. Both the highest alkaloid diversity and the most potent inhibitory activity (IC50 12.94 microg/ml) were obtained in extracts from the bulbs of G. rizehensis collected during the fruiting period.
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (Lowry)
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from human erythrocytes, buffered aqueous solution, ≥500 units/mg protein (BCA)