Merck
  • Agalsidase benefits renal histology in young patients with Fabry disease.
Agalsidase benefits renal histology in young patients with Fabry disease.
Journal of the American Society of Nephrology : JASN (2013-01-01)
Camilla Tøndel, Leif Bostad, Kristin Kampevold Larsen, Asle Hirth, Bjørn Egil Vikse, Gunnar Houge, Einar Svarstad
ABSTRACT
The effect of early-onset enzyme replacement therapy on renal morphologic features in Fabry disease is largely unknown. Here, we evaluated the effect of 5 years of treatment with agalsidase alfa or agalsidase beta in 12 consecutive patients age 7-33 years (median age, 16.5 years). We performed renal biopsies at baseline and after 5 years of enzyme replacement therapy; 7 patients had additional biopsies after 1 and 3 years. After a median of 65 months, biopsy findings from all patients showed total clearance of glomerular endothelial and mesangial cell inclusions, and findings from 2 patients showed complete clearance of inclusions from epithelial cells of the distal tubule. The 4 patients who received the highest dose of agalsidase exhibited substantial clearance of podocyte inclusions, and the youngest patient had nearly complete clearance of these inclusions. Linear regression analysis showed a highly significant correlation between podocyte globotriaocylceramide clearance and cumulative agalsidase dose (r=0.804; P=0.002). Microalbuminuria normalized in five patients. In summary, long-term enzyme replacement therapy in young patients can result in complete globotriaocylceramide clearance of mesangial and glomerular endothelial cells across all dosage regimens, and clearance of podocyte inclusions is dose-dependent.
Sigma-Aldrich
α-Galactosidase from green coffee beans, ammonium sulfate suspension, ≥9 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
α-Galactosidase, positionally specific from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution
Pricing and availability is not currently available.