Merck
  • An enzymatic method to determine γ-hydroxybutyric acid in serum and urine.
An enzymatic method to determine γ-hydroxybutyric acid in serum and urine.
Therapeutic drug monitoring (2011-11-23)
Lara Hasan, Thomas M Jermann, Jakob M Weber, Lars Abrahamsson, Michel-Angelo Sciotti, Michael Böttcher, Wolfgang Jöchle, Daniel Gygax, André Scholer
ABSTRACT
Gamma-hydroxybutyric acid (GHB) has become one of the most dangerous illicit drugs of abuse today. It is used as a recreational and date rape drug because of its depressant effect on the central nervous system, which may cause euphoria, amnesia, respiratory arrest, and coma. There is an urgent need for a simple, easy-to-use assay for GHB determination in urine and blood. In this article, a rapid enzymatic assay adapted to clinical chemistry analyzers for the detection of GHB is presented. The described GHB enzymatic assay is based on a recombinant GHB dehydrogenase. The full validation of the assay was performed on a Konelab 30 analyzer (Thermo Fisher Scientific). The analytical sensitivity was <1.5 mg/L, whereas the functional sensitivity was 4.5 mg/L in serum and 2.8 mg/L in urine. The total imprecision coefficient of variation (CV) was <9.8% in serum and <7.9% in urine. The within-run imprecision showed a CV of <3.8% in serum and <4.6% in urine. The assay was linear within the range 5-250 mg/L. Mean recoveries were 109% in serum and 105% in urine. No cross-reactivity was observed for tested GHB analogues and precursors. Comparison of GHB-positive samples showed an excellent correlation with ion chromatography, gas chromatography-mass spectrometry, and liquid chromatography associated to tandem mass spectrometry. Except for ethanol, no substantial interference from serum constituents and some drugs was observed. This automated GHB assay is fully quantitative and allows the accurate measurement of GHB in serum and urine. It can be used as a rapid screening assay for the determination of GHB in intoxicated or overdosed patients.