Merck
Fluorescence quenching by TEMPO: a sub-30 A single-molecule ruler.
Biophysical journal (2005-10-04)
Peizhi Zhu, Jean-Pierre Clamme, Ashok A Deniz
ABSTRACT
A series of DNA molecules labeled with 5-carboxytetramethylrhodamine (5-TAMRA) and the small nitroxide radical TEMPO were synthesized and tested to investigate whether the intramolecular quenching efficiency can be used to measure short intramolecular distances in small ensemble and single-molecule experiments. In combination with distance calculations using molecular mechanics modeling, the experimental results from steady-state ensemble fluorescence and fluorescence correlation spectroscopy measurements both show an exponential decrease in the quenching rate constant with the dye-quencher distance in the 10-30 A range. The results demonstrate that TEMPO-5-TAMRA fluorescence quenching is a promising method to measure short distance changes within single biomolecules.
Sigma-Aldrich
5-Carboxy-tetramethylrhodamine N-succinimidyl ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
5-Carboxytetramethylrhodamine