Merck
  • Interferon and cell division. I. Inhibition of the multiplication of mouse leukemia L 1210 cells in vitro by interferon preparations.
Interferon and cell division. I. Inhibition of the multiplication of mouse leukemia L 1210 cells in vitro by interferon preparations.
Proceedings of the National Academy of Sciences of the United States of America (1970-08-01)
I Gresser, D Brouty-Boyé, M T Thomas, A Macieira-Coelho
ABSTRACT
Mouse interferon preparations inhibited the multiplication of mouse leukemia L 1210 cells in stationary suspension cultures. The degree of inhibition was found to correlate with the antiviral titer of the interferon preparations. The factor(s) responsible for inhibition of L 1210 cell multiplication could not be dissociated from interferon by standard physicochemical means.