Merck
  • Fibroblast-Specific Depletion of Human Antigen R Alleviates Myocardial Fibrosis Induced by Cardiac Stress.
Fibroblast-Specific Depletion of Human Antigen R Alleviates Myocardial Fibrosis Induced by Cardiac Stress.
JACC. Basic to translational science (2024-07-29)
Mallikarjun Patil, Sarojini Singh, Praveen Kumar Dubey, Sultan Tousif, Prachi Umbarkar, Qinkun Zhang, Hind Lal, Mary Kathryn Sewell-Loftin, Channakeshava Sokke Umeshappa, Yohannes T Ghebre, Steven Pogwizd, Jianyi Zhang, Prasanna Krishnamurthy
ABSTRACT
Cardiac fibrosis can be mitigated by limiting fibroblast-to-myofibroblast differentiation and proliferation. Human antigen R (HuR) modulates messenger RNA stability and expression of multiple genes. However, the direct role of cardiac myofibroblast HuR is unknown. Myofibroblast-specific deletion of HuR limited cardiac fibrosis and preserved cardiac functions in pressure overload injury. Knockdown of HuR in transforming growth factor-β1-treated cardiac fibroblasts suppressed myofibroblast differentiation and proliferation. HuR deletion abrogated the expression and messenger RNA stability of cyclins D1 and A2, suggesting a potential mechanism by which HuR promotes myofibroblast proliferation. Overall, these data suggest that inhibition of HuR could be a potential therapeutic approach to limit cardiac fibrosis.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%