Merck
  • Fisetin inhibits Salmonella Typhimurium type III secretion system regulator HilD and reduces pathology in vivo.
Fisetin inhibits Salmonella Typhimurium type III secretion system regulator HilD and reduces pathology in vivo.
Microbiology spectrum (2023-12-11)
Siqi Li, Hongtao Liu, Jingyan Shu, Quanshun Li, Yuan Liu, Haihua Feng, Jianfeng Wang, Xuming Deng, Yong Zhang, Zhimin Guo, Jiazhang Qiu
ABSTRACT
Salmonella spp. remains a major worldwide health concern that causes significant morbidity and mortality in both humans and animals. The spread of antimicrobial resistant strains has declined the efficacy of conventional chemotherapy. Thus, novel anti-infection drugs or strategies are needed. Anti-virulence strategy represents one of the promising means for the treatment of bacterial infections. In this study, we found that the natural compound fisetin could inhibit Salmonella invasion of host cells by targeting SPI-1 regulation. Fisetin treatment impaired the interaction of the regulatory protein HilD with the promoters of its target genes, thereby suppressing the expression of T3SS-1 effectors as well as structural proteins. Moreover, fisetin treatment could reduce pathology in the Salmonella murine infection model. Collectively, our results suggest that fisetin may serve as a promising lead compound for the development of anti-Salmonella drugs.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-ICDH, serum, from rabbit
Pricing and availability is not currently available.