Merck
  • Progressive alterations in polysomal architecture and activation of ribosome stalling relief factors in a mouse model of Huntington's disease.
Progressive alterations in polysomal architecture and activation of ribosome stalling relief factors in a mouse model of Huntington's disease.
Neurobiology of disease (2024-04-03)
Eva Martin-Solana, Irene Diaz-Lopez, Yamina Mohamedi, Ivan Ventoso, Jose-Jesus Fernandez, Maria Rosario Fernandez-Fernandez
ABSTRACT
Given their highly polarized morphology and functional singularity, neurons require precise spatial and temporal control of protein synthesis. Alterations in protein translation have been implicated in the development and progression of a wide range of neurological and neurodegenerative disorders, including Huntington's disease (HD). In this study we examined the architecture of polysomes in their native brain context in striatal tissue from the zQ175 knock-in mouse model of HD. We performed 3D electron tomography of high-pressure frozen and freeze-substituted striatal tissue from HD models and corresponding controls at different ages. Electron tomography results revealed progressive remodelling towards a more compacted polysomal architecture in the mouse model, an effect that coincided with the emergence and progression of HD related symptoms. The aberrant polysomal architecture is compatible with ribosome stalling phenomena. In fact, we also detected in the zQ175 model an increase in the striatal expression of the stalling relief factor EIF5A2 and an increase in the accumulation of eIF5A1, eIF5A2 and hypusinated eIF5A1, the active form of eIF5A1. Polysomal sedimentation gradients showed differences in the relative accumulation of 40S ribosomal subunits and in polysomal distribution in striatal samples of the zQ175 model. These findings indicate that changes in the architecture of the protein synthesis machinery may underlie translational alterations associated with HD, opening new avenues for understanding the progression of the disease.
Sigma-Aldrich
Monoclonal Anti-EIF5A antibody produced in mouse, clone 8C1, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.