Merck
  • Astrocyte ensheathment of calyx-forming axons of the auditory brainstem precedes accelerated expression of myelin genes and myelination by oligodendrocytes.
Astrocyte ensheathment of calyx-forming axons of the auditory brainstem precedes accelerated expression of myelin genes and myelination by oligodendrocytes.
The Journal of comparative neurology (2023-11-02)
Daniel T Heller, Douglas R Kolson, Ashley N Brandebura, Emily M Amick, Jun Wan, Jad Ramadan, Paul S Holcomb, Sheng Liu, Thomas J Deerinck, Mark H Ellisman, Jiang Qian, Peter H Mathers, George A Spirou
ABSTRACT
Early postnatal brain development involves complex interactions among maturing neurons and glial cells that drive tissue organization. We previously analyzed gene expression in tissue from the mouse medial nucleus of the trapezoid body (MNTB) during the first postnatal week to study changes that surround rapid growth of the large calyx of Held (CH) nerve terminal. Here, we present genes that show significant changes in gene expression level during the second postnatal week, a developmental timeframe that brackets the onset of airborne sound stimulation and the early stages of myelination. Gene Ontology analysis revealed that many of these genes are related to the myelination process. Further investigation of these genes using a previously published cell type-specific bulk RNA-Seq data set in cortex and our own single-cell RNA-Seq data set in the MNTB revealed enrichment of these genes in the oligodendrocyte lineage (OL) cells. Combining the postnatal day (P)6-P14 microarray gene expression data with the previously published P0-P6 data provided fine temporal resolution to investigate the initiation and subsequent waves of gene expression related to OL cell maturation and the process of myelination. Many genes showed increasing expression levels between P2 and P6 in patterns that reflect OL cell maturation. Correspondingly, the first myelin proteins were detected by P4. Using a complementary, developmental series of electron microscopy 3D image volumes, we analyzed the temporal progression of axon wrapping and myelination in the MNTB. By employing a combination of established ultrastructural criteria to classify reconstructed early postnatal glial cells in the 3D volumes, we demonstrated for the first time that astrocytes within the mouse MNTB extensively wrap the axons of the growing CH terminal prior to OL cell wrapping and compaction of myelin. Our data revealed significant expression of several myelin genes and enrichment of multiple genes associated with lipid metabolism in astrocytes, which may subserve axon wrapping in addition to myelin formation. The transition from axon wrapping by astrocytes to OL cells occurs rapidly between P4 and P9 and identifies a potential new role of astrocytes in priming calyceal axons for subsequent myelination.
Sigma-Aldrich
Anti-Aldh1L1 Antibody, clone N103/39, clone N103/39, from mouse
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neurofilament H Antibody, Chemicon®, from chicken