Merck
Regulation of folate transport at the mouse arachnoid barrier.
Fluids and barriers of the CNS (2024-08-28)
Vishal Sangha, Sara Aboulhassane, Reina Bendayan
ABSTRACT
Folates are a family of B9 vitamins essential for normal growth and development in the central nervous system (CNS). Transport of folates is mediated by three major transport proteins: folate receptor alpha (FRα), proton-coupled folate transporter (PCFT), and reduced folate carrier (RFC). Brain folate uptake occurs at the choroid plexus (CP) epithelium through coordinated actions of FRα and PCFT, or directly into brain parenchyma at the vascular blood-brain barrier (BBB), mediated by RFC. Impaired folate transport can occur due to loss of function mutations in FRα or PCFT, resulting in suboptimal CSF folate levels. Our previous reports have demonstrated RFC upregulation by nuclear respiratory factor-1 (NRF-1) once activated by the natural compound pyrroloquinoline quinone (PQQ). More recently, we have identified folate transporter localization at the arachnoid barrier (AB). The purpose of the present study was to further characterize folate transporters localization and function in AB cells, as well as their regulation by NRF-1/PGC-1α signaling and folate deficiency. In immortalized mouse AB cells, polarized localization of RFC and PCFT was assessed by immunocytochemical analysis, with RFC and PCFT functionality examined with transport assays. The effects of PQQ treatment on changes in RFC functional expression were also investigated. Mouse AB cells grown in folate-deficient conditions were assessed for changes in gene expression of the folate transporters, and other key transporters and tight junction proteins. Immunocytochemical analysis revealed apical localization of RFC at the mouse AB epithelium, with PCFT localized on the basolateral side and within intracellular compartments. PQQ led to significant increases in RFC functional expression, mediated by activation of the NRF-1/PGC-1α signalling cascade. Folate deficiency led to significant increases in expression of RFC, MRP3, P-gp, GLUT1 and the tight junction protein claudin-5. These results uncover the polarized expression of RFC and PCFT at the AB, with induction of RFC functional expression by activation of the NRF-1/PGC-1α signalling pathway and folate deficiency. These results suggest that the AB may contribute to the flow of folates into the CSF, representing an additional pathway when folate transport at the CP is impaired.
Sigma-Aldrich
Anti-SLC46A1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-SLC19A1 antibody produced in rabbit, IgG fraction of antiserum