MilliporeSigma
  • Pandemic-associated pernio harbors footprints of an abortive SARS-CoV-2 infection.
Pandemic-associated pernio harbors footprints of an abortive SARS-CoV-2 infection.
iScience (2024-08-19)
Lisa M Arkin, Ana C Costa-da-Silva, Justin Frere, Ashley Ng, Rubina Sharma, John J Moon, Hailey E Bussan, Clara H Kim, Ayesha Javaid, Olivia R Steidl, Ahmad Yatim, Fanny Saidoune, Michel Gilliet, Joe T Nguyen, Aman Nihal, George Luong, Meaghan Kenfield, Lucia Carrau, Jennifer M Tran, Molly A Hinshaw, Erin G Brooks, Jose M Ayuso, David H O'Connor, Jean-Laurent Casanova, Edward W Cowen, Beth A Drolet, Anne Marie Singh, Benjamin tenOever, Jacqueline W Mays
ABSTRACT
Elevated pernio incidence was observed during the COVID-19 pandemic. This prospective study enrolled subjects with pandemic-associated pernio in Wisconsin and Switzerland. Because pernio is a cutaneous manifestation of the interferonopathies, and type I interferon (IFN-I) immunity is critical to COVID-19 recovery, we tested the hypothesis that severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)-mediated IFN-I signaling might underlie some pernio cases. Tissue-level IFN-I activity and plasmacytoid dendritic cell infiltrates were demonstrated in 100% of the Wisconsin cases. Across both cohorts, sparse SARS-CoV-2 RNA was captured in 25% (6/22) of biopsies, all with high inflammation. Affected patients lacked adaptive immunity to SARS-CoV-2. A hamster model of intranasal SARS-CoV-2 infection was used as a proof-of-principle experiment: RNA was detected in lungs and toes with IFN-I activity at both the sites, while replicating virus was found only in the lung. These data support a viral trigger for some pernio cases, where sustained local IFN-I activity can be triggered in the absence of seroconversion.