Merck
  • Discovery and Characterization of BAY-805, a Potent and Selective Inhibitor of Ubiquitin-Specific Protease USP21.
Discovery and Characterization of BAY-805, a Potent and Selective Inhibitor of Ubiquitin-Specific Protease USP21.
Journal of medicinal chemistry (2023-02-22)
Fabian Göricke, Victoria Vu, Leanna Smith, Ulrike Scheib, Raphael Böhm, Namik Akkilic, Gerd Wohlfahrt, Jörg Weiske, Ulf Bömer, Krzysztof Brzezinka, Niels Lindner, Philip Lienau, Stefan Gradl, Hartmut Beck, Peter J Brown, Vijayaratnam Santhakumar, Masoud Vedadi, Dalia Barsyte-Lovejoy, Cheryl H Arrowsmith, Norbert Schmees, Kirstin Petersen
ABSTRACT
USP21 belongs to the ubiquitin-specific protease (USP) subfamily of deubiquitinating enzymes (DUBs). Due to its relevance in tumor development and growth, USP21 has been reported as a promising novel therapeutic target for cancer treatment. Herein, we present the discovery of the first highly potent and selective USP21 inhibitor. Following high-throughput screening and subsequent structure-based optimization, we identified BAY-805 to be a non-covalent inhibitor with low nanomolar affinity for USP21 and high selectivity over other DUB targets as well as kinases, proteases, and other common off-targets. Furthermore, surface plasmon resonance (SPR) and cellular thermal shift assays (CETSA) demonstrated high-affinity target engagement of BAY-805, resulting in strong NF-κB activation in a cell-based reporter assay. To the best of our knowledge, BAY-805 is the first potent and selective USP21 inhibitor and represents a valuable high-quality in vitro chemical probe to further explore the complex biology of USP21.
Sigma-Aldrich
BAY-805, ≥98% (HPLC)