Merck
  • A mouse model for monitoring calpain activity under physiological and pathological conditions.
A mouse model for monitoring calpain activity under physiological and pathological conditions.
The Journal of biological chemistry (2006-10-24)
Marc Bartoli, Nathalie Bourg, Daniel Stockholm, Fabrice Raynaud, Antony Delevacque, Yang Han, Perrine Borel, Kenza Seddik, Nasser Armande, Isabelle Richard
ABSTRACT
Calpains are Ca(2+)-dependent cysteine proteases known to be important for the regulation of cell functions and which aberrant activation causes cell death in a number of degenerative disorders. To provide a tool for monitoring the status of calpain activity in vivo under physiological and pathological conditions, we created a mouse model that expresses ubiquitously a fluorescent reporter consisting of eCFP and eYFP separated by a linker cleavable by the ubiquitous calpains. We named this mouse CAFI for calpain activity monitored by FRET imaging. Our validation studies demonstrated that the level of calpain activity correlates with a decrease in FRET (fluorescence resonance energy transfer) between the two fluorescent proteins. Using this model, we observed a small level of activity after denervation and fasting, a high level of activity during muscle regeneration and ischemia, and local activity in damaged myofibers after exercise. Finally, we crossed the CAFI mouse with the alpha-sarcoglycan-deficient model, demonstrating an increase of calpain activity at the steady state. Altogether, our results present evidence that CAFI mice could be a valuable tool in which to follow calpain activity at physiological levels and in disease states.