Merck
  • Effect of Collagen and GelMA on Preservation of the Costal Chondrocytes' Phenotype in a Scaffold in vivo.
Effect of Collagen and GelMA on Preservation of the Costal Chondrocytes' Phenotype in a Scaffold in vivo.
Sovremennye tekhnologii v meditsine (2023-06-30)
E V Isaeva, A A Kisel, E E Beketov, G A Demyashkin, N D Yakovleva, T S Lagoda, N V Arguchinskaya, D S Baranovsky, S A Ivanov, P V Shegay, A D Kaprin
ABSTRACT
The aim of the study was to compare type I collagen-based and methacryloyl gelatin-based (GelMA) hydrogels by their ability to form hyaline cartilage in animals after subcutaneous implantation of scaffolds. Chondrocytes were isolated from the costal cartilage of newborn rats using 0.15% collagenase solution in DMEM. The cells was characterized by glycosaminoglycan staining with alcian blue. Chondrocyte scaffolds were obtained from 4% type I porcine atelocollagen and 10% GelMA by micromolding and then implanted subcutaneously into the withers of two groups of Wistar rats. Histological and immunohistochemical studies were performed on days 12 and 26 after implantation. Tissue samples were stained with hematoxylin and eosin, alcian blue; type I and type II collagens were identified by the corresponding antibodies. The implanted scaffolds induced a moderate inflammatory response in both groups when implanted in animals. By day 26 after implantation, both collagen and GelMA had almost completely resorbed. Cartilage tissue formation was observed in both animal groups. The newly formed tissue was stained intensively with alcian blue, and the cells were positive for both types of collagen. Cartilage tissue was formed among muscle fibers. The ability of collagen type I and GelMA hydrogels to form hyaline cartilage in animals after subcutaneous implantation of scaffolds was studied. Both collagen and GelMA contributed to formation of hyaline-like cartilage tissue type in animals, but the chondrocyte phenotype is characterized as mixed. Additional detailed studies of possible mechanisms of chondrogenesis under the influence of each of the hydrogels are needed.
Sigma-Aldrich
Anti-Collagen II antibody produced in rabbit, affinity isolated antibody