Merck
Pharmacological effects and mechanisms of tannic acid.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2022-08-28)
Wang Jing, Chen Xiaolan, Chen Yu, Qin Feng, Yang Haifeng
ABSTRACT
In recent years, increasing attention has been paid to the pharmacological efficacy of tannins. Tannic acid (TA), the simplest hydrolysable tannin that has been approved by the FDA as a safe food additive, is one of the most important components of these traditional medicines. Studies have shown that TA displays a wide range of pharmacological activities, such as anti-inflammatory, neuroprotective, antitumor, cardioprotective, and anti-pathogenic effects. Here, we summarize the known pharmacological effects and associated mechanisms of TA. We focus on the effect and mechanism of TA in various animal models of inflammatory disease and organ, brain, and cardiovascular injury. Moreover, we discuss the possible molecular targets and signaling pathways of TA, in addition to the pharmacological effects of TA-based nanoparticles and TA in combination with chemotherapeutic drugs.