Merck
  • Pharmacological Activity of Eriodictyol: The Major Natural Polyphenolic Flavanone.
Pharmacological Activity of Eriodictyol: The Major Natural Polyphenolic Flavanone.
Evidence-based complementary and alternative medicine : eCAM (2021-01-09)
Zirong Deng, Sabba Hassan, Muhammad Rafiq, Hongshui Li, Yang He, Yi Cai, Xi Kang, Zhaoguo Liu, Tingdong Yan
ABSTRACT
Eriodictyol is a flavonoid that belongs to a subclass of flavanones and is widespread in citrus fruits, vegetables, and medicinally important plants. Eriodictyol has been anticipated to explain the method of its activity via multiple cellular signaling cascades. Eriodictyol is an effective natural drug source to maintain higher health standards due to its excellent therapeutic roles in neuroprotection, cardioprotective activity, hepatoprotective activity, antidiabetes and obesity, and skin protection and having highly analgesic, antioxidant, and anti-inflammatory effects, antipyretic and antinociceptive actions, antitumor activity, and much more. This review aims to highlight the modes of action of eriodictyol against various diseases via multiple cellular signaling pathways.
Sigma-Aldrich
Eriodictyol, ≥95.0% (HPLC)
Pricing and availability is not currently available.