- Cathepsin L inhibition prevents the cleavage of multiple nuclear proteins upon lysis of quiescent human cells.
Cathepsin L inhibition prevents the cleavage of multiple nuclear proteins upon lysis of quiescent human cells.
microPublication biology (2023-01-07)
Prashant Gaikwad, Michael G Kemp
PMID36606083
ABSTRACT
Several studies have indicated a role for cathepsin L (CTSL) proteolytic activity in the nucleus under distinct cellular conditions, including during differentiation, senescence, and quiescence. Here we show that addition of CTSL inhibitors to a cell lysis buffer prevents the cleavage of several nuclear proteins during the lysis of quiescent human cells, including proteins previously thought to have functional relevance in other cell and tissue contexts. These findings suggest that care should be taken to use CTSL inhibitors when lysing cells and tissues containing high levels of CTSL protein to differentiate proteolysis that occurs in vivo versus artifactually in vitro.
Sigma-Aldrich
Cathepsin L Inhibitor II, The Cathepsin L Inhibitor II controls the biological activity of Cathepsin L. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.