Merck
  • Transcription factor immunohistochemistry in the diagnosis of pituitary tumours.
Transcription factor immunohistochemistry in the diagnosis of pituitary tumours.
European journal of endocrinology (2021-04-15)
Nèle F Lenders, Adam C Wilkinson, Stephen J Wong, Tint T Shein, Richard J Harvey, Warrick J Inder, Peter E Earls, Ann I McCormack
ABSTRACT
The clinical utility and prognostic value of WHO 2017 lineage-based classification of pituitary tumours have not been assessed. This study aimed to (1) determine the clinical utility of transcription factor analysis for classification of pituitary tumours and (2) determine the prognostic value of improved lineage-based classification of pituitary tumours. This was a retrospective evaluation of patients who underwent surgical resection of pituitary tumours at St Vincent's Public and Private Hospitals, Sydney, Australia between 1990 and 2016. Included patients were at least 18 years of age and had complete histopathological data, forming the 'histological cohort'. Patients with at least 12 months of post-surgical follow-up were included in the subgroup 'clinical cohort'. The diagnostic efficacy of transcription factor immunohistochemistry in conjunction with hormone immunohistochemistry was compared with hormone immunohistochemistry alone. The prognostic value of identifying 'higher-risk' histological subtypes was assessed. There were 171 patient tumour samples analyzed in the histological cohort. Of these, there were 95 patients forming the clinical cohort. Subtype diagnosis was changed in 20/171 (12%) of tumours. Within the clinical cohort, there were 21/95 (22%) patients identified with higher-risk histological subtype tumours. These were associated with tumour invasiveness (P = 0.050), early recurrence (12-24 months, P = 0.013), shorter median time to recurrence (49 (IQR: 22.5-73.0) vs 15 (IQR: 12.0-25.0) months, P = 0.005) and reduced recurrence-free survival (P = 0.031). Application of transcription factor analysis, in addition to hormone immunohistochemistry, allows for refined pituitary tumour classification and may facilitate an improved approach to prognostication.
Sigma-Aldrich
Monoclonal Anti-TBX19 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL6251, purified immunoglobulin, buffered aqueous glycerol solution
Pricing and availability is not currently available.