Merck
  • Exploration of the cysteine reactivity of human inducible Hsp70 and cognate Hsc70.
Exploration of the cysteine reactivity of human inducible Hsp70 and cognate Hsc70.
The Journal of biological chemistry (2022-11-22)
Zhouping Hong, Weibin Gong, Jie Yang, Sainan Li, Zhenyan Liu, Sarah Perrett, Hong Zhang
ABSTRACT
Hsp70s are multifunctional proteins and serve as the central hub of the protein quality control network. Hsp70s are also related to a number of diseases and have been established as drug targets. Human HspA1A (hHsp70) and HspA8 (hHsc70) are the major cytosolic Hsp70s, and they have both overlapping and distinct functions. hHsp70 contains five cysteine residues, and hHsc70 contains four cysteine residues. Previous studies have shown these cysteine residues can undergo different cysteine modifications such as oxidation or reaction with electrophiles to regulate their function, and hHsp70 and hHsc70 have different cysteine reactivity. To address the mechanism of the differences in cysteine reactivity between hHsp70 and hHsc70, we studied the factors that determine this reactivity by Ellman assay for the quantification of accessible free thiols and NMR analysis for the assessment of structural dynamics. We found the lower cysteine reactivity of hHsc70 is probably due to its lower structural dynamics and the stronger inhibition effect of interaction between the α-helical lid subdomain of the substrate-binding domain (SBDα) and the β-sheet substrate-binding subdomain (SBDβ) on cysteine reactivity of hHsc70. We determined that Gly557 in hHsp70 contributes significantly to the higher structural dynamics and cysteine reactivity of hHsp70 SBDα. Exploring the cysteine reactivity of hHsp70 and hHsc70 facilitates an understanding of the effects of redox reactions and electrophiles on their chaperone activity and regulation mechanisms, and how these differences allow them to undertake distinct cellular roles.
Sigma-Aldrich
Anti-Glutathione Antibody, detects both GSH & GSSG, Chemicon®, from rabbit
Pricing and availability is not currently available.