Merck
  • Examination of Immunogenic Properties of Recombinant Antigens Based on p22 Protein from African Swine Fever Virus.
Examination of Immunogenic Properties of Recombinant Antigens Based on p22 Protein from African Swine Fever Virus.
Journal of veterinary research (2022-11-10)
Carlos Díaz, Jiří Salát, Dagmar Břínek Kolařová, Vladimír Celer, Ivo Frébort
ABSTRACT
The single member of the Asfarviridae family is African swine fever virus (ASFV). This double-stranded DNA virus infects wild and farmed swine and loses the pig industry large sums of money. An inner envelope, capsid, and outer envelope are parts of the ASFV particle containing structural proteins playing different roles in the process of infection or host immune defence evasion. When expressed by the baculovirus system, the p22 protein from the inner envelope was found to induce partial protection against a virulent virus strain. This study aimed to express a part of this protein in a different system and evaluate its immunogenicity. We designed two proteins, the extracellular (C terminal) part of the p22 protein (p22Ct) and its fusion with the heat-labile enterotoxin B subunit from Escherichia coli (LTB-p22Ct), which is supposed to be a potent enhancer of the immune response. Both proteins were produced in the E. coli expression system and subsequently used for mice immunisation to analyse their safety and immunogenicity. The protein fused with LTB did not show the expected adjuvant properties and did not prove safe, because abscess formation was observed after immunisation. In contrast, immunisation with the p22Ct protein alone induced a higher antibody titre but caused no adverse symptoms. These results show the high potential of the p22Ct region as an immunogenic protein for ASFV serological detection purposes.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Pig IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes, ready to use solution