Merck
  • Corticotropin-releasing factor neurogenesis during midlife development in salmon: genetic, environmental and thyroid hormone regulation.
Corticotropin-releasing factor neurogenesis during midlife development in salmon: genetic, environmental and thyroid hormone regulation.
Journal of neuroendocrinology (2011-05-20)
L O E Ebbesson, T O Nilsen, J V Helvik, V Tronci, S O Stefansson
ABSTRACT
Salmon parr-smolt transformation (smoltification) is a mid-life transitional stage between life in freshwater and seawater that entails a wide range of neural, endocrine and physiological modifications. In salmon, the neuroendocrine corticotropin-releasing factor (CRF) system regulates pituitary adrenocorticotrophic hormone and thyrotrophin release. Four experimental groups of Atlantic salmon, Salmo salar, were used to investigated CRF neurogenesis and its regulation during smoltification. We compared: (i) developmental stages (parr and early-smolt) in anadromous controls; (ii) a developmentally arrested model: anadromous reared under continuous light (LL) with anadromous controls; (iii) a natural hypoendocrine/incomplete smolt development salmon model (landlocked) with anadromous controls; and (iv) landlocked treated with thyroxine to anadromous control smolt levels. CRF neurogenesis between groups was studied with bromodeoxyuradine (BrdU) incorporation followed by double-labelling CRF and BrdU immunhistochemistry. The rate of CRF neurogenesis in the preoptic area (POA) increased from parr to early-smolts in anadromous salmon. By contrast, neurogenesis was inhibited in the LL group and reduced in the landlocked salmon. The administration of thyroxine in landlocked salmon to match anadromous levels increased the rate of CRF neurogenesis to anadromous levels. In conclusion, newly-formed CRF cells in the POA during smoltification are associated with increased retinal innervation to the POA and endocrine responsiveness to increased photoperiod. Both genetic and environmental factors influence the degree of salmon brain development. Thyroid hormones increase CRF neurogenesis during this critical period of development in salmon. We hypothesise that a positive-feedback of thyroid hormones on CRF neurogenesis may be an important event in reaching the developmental climax during critical periods.
Sigma-Aldrich
Corticotropin Releasing Factor sheep, ≥95% (HPLC)