Merck
  • Blockage of PPARγ T166 phosphorylation enhances the inducibility of beige adipocytes and improves metabolic dysfunctions.
Blockage of PPARγ T166 phosphorylation enhances the inducibility of beige adipocytes and improves metabolic dysfunctions.
Cell death and differentiation (2022-11-05)
Nanfei Yang, Yuxin Wang, Qiang Tian, Qiuping Wang, Yan Lu, Luchen Sun, Sijie Wang, Yuncheng Bei, Jianguo Ji, Hu Zhou, Wei Yang, Pengju Yao, Wenyuan Zhu, Lingyun Sun, Zhifeng Huang, Xiaokun Li, Pingping Shen
ABSTRACT
Beige adipocytes in mammalian white adipose tissue (WAT) can reinforce fat catabolism and energy expenditure. Promoting beige adipocyte biogenesis is a tantalizing tactic for combating obesity and its associated metabolic disorders. Here, we report that a previously unidentified phosphorylation pattern (Thr166) in the DNA-binding domain of PPARγ regulates the inducibility of beige adipocytes. This unique posttranslational modification (PTM) pattern influences allosteric communication between PPARγ and DNA or coactivators, which impedes the PPARγ-mediated transactivation of beige cell-related gene expression in WAT. The genetic mutation mimicking T166 phosphorylation (p-T166) hinders the inducibility of beige adipocytes. In contrast, genetic or chemical intervention in this PTM pattern favors beige cell formation. Moreover, inhibition of p-T166 attenuates metabolic dysfunction in obese mice. Our results uncover a mechanism involved in beige cell fate determination. Moreover, our discoveries provide a promising strategy for guiding the development of novel PPARγ agonists for the treatment of obesity and related metabolic disorders.
Sigma-Aldrich
Anti-phospho-Histone H1 Antibody, clone 12D11, culture supernatant, clone 12D11, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Histone H1 Protein, 20 mg, Purified bovine Histone H1 protein for use as a substrate in histone modification assays (HAT, HDAC, DNMT) and chromatin assembly studies.
Pricing and availability is not currently available.