Merck
  • 2-Aminothiophene derivatives as a new class of positive allosteric modulators of glucagon-like peptide 1 receptor.
2-Aminothiophene derivatives as a new class of positive allosteric modulators of glucagon-like peptide 1 receptor.
Chemical biology & drug design (2022-03-22)
Tejashree Redij, James A McKee, Phu Do, Jeffrey A Campbell, Jian Ma, Zhiyu Li, Nicholas Miller, Chananchida Srikanlaya, Dianzheng Zhang, Xianxin Hua, Zhijun Li
ABSTRACT
We report the discovery of two new 2-aminothiophene based small molecule positive allosteric modulators (PAMs) of glucagon-like peptide 1 receptor (GLP-1R) for the treatment of type 2 diabetes. One of the chemotypes, (S-1), has a molecular weight of 239 g/mol, the smallest molecule among all reported GLP-1R PAMs. When combined with GLP-1 peptide, S-1 increased the GLP-1R activity in a dose-dependent manner in a cell-based assay. When combined with the peptide agonist of vasoactive intestinal polypeptide receptor 1 (VIPR1), S-1 showed no specific activity on VIPR1, another class B GPCR present in the same HEK293-CREB cell line. Insulin secretion studies found S-1 combined with GLP-1 increased insulin secretion by 1.5-fold at 5 μM. In a mechanistic study, evidence is provided that the synergistic effect of S-1 with GLP-1 may be partly due to the enhanced impact on CREB based phosphorylation. Given the favorable profile of these chemotypes, the work reported herein suggests that 2-aminothiophene derivatives are a new and promising class of GLP-1R PAMs.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
[Ac-Tyr1,D-Phe2]-VIP Antagonist-GRF 1-29
Pricing and availability is not currently available.