Merck
  • In vivo transcriptomic profiling using cell encapsulation identifies effector pathways of systemic aging.
In vivo transcriptomic profiling using cell encapsulation identifies effector pathways of systemic aging.
eLife (2022-03-05)
Omid Mashinchian, Xiaotong Hong, Joris Michaud, Eugenia Migliavacca, Gregory Lefebvre, Christophe Boss, Filippo De Franceschi, Emmeran Le Moal, Jasmin Collerette-Tremblay, Joan Isern, Sylviane Metairon, Frederic Raymond, Patrick Descombes, Nicolas Bouche, Pura Muñoz-Cánoves, Jerome N Feige, C Florian Bentzinger
ABSTRACT
Sustained exposure to a young systemic environment rejuvenates aged organisms and promotes cellular function. However, due to the intrinsic complexity of tissues it remains challenging to pinpoint niche-independent effects of circulating factors on specific cell populations. Here, we describe a method for the encapsulation of human and mouse skeletal muscle progenitors in diffusible polyethersulfone hollow fiber capsules that can be used to profile systemic aging in vivo independent of heterogeneous short-range tissue interactions. We observed that circulating long-range signaling factors in the old systemic environment lead to an activation of Myc and E2F transcription factors, induce senescence, and suppress myogenic differentiation. Importantly, in vitro profiling using young and old serum in 2D culture does not capture all pathways deregulated in encapsulated cells in aged mice. Thus, in vivo transcriptomic profiling using cell encapsulation allows for the characterization of effector pathways of systemic aging with unparalleled accuracy.
Sigma-Aldrich
TrueGel3D Hydrogel Kits, SLO-DEXTRAN, allow cell recovery, CD cell-degradable crosslinker
Pricing and availability is not currently available.
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Pricing and availability is not currently available.