- ADA2b and GCN5 Affect Cytokinin Signaling by Modulating Histone Acetylation and Gene Expression during Root Growth of Arabidopsis thaliana.
ADA2b and GCN5 Affect Cytokinin Signaling by Modulating Histone Acetylation and Gene Expression during Root Growth of Arabidopsis thaliana.
In Arabidopsis thaliana, the histone acetyltransferase GCN5 and the associated coactivator ADA2b regulate root growth and affect gene expression. The cytokinin signaling reporter TCS::GFP was introduced into gcn5-1, ada2b-1, and ada2a-2, as well as the ada2a-2ada2b-1 mutants. The early root growth (4 to 7 days post-germination) was analyzed using cellular and molecular approaches. TCS signal accumulated from the fourth to seventh days of root growth in the wild-type columella cells. In contrast, ada2b-1 and gcn5-1 and ada2a-2ada2b-1 double mutants displayed reduced TCS expression relative to wild type. Gene expression analysis showed that genes associated with cytokinin homeostasis were downregulated in the roots of gcn5-1 and ada2b-1 mutants compared to wild-type plants. H3K14 acetylation was affected in the promoters of cytokinin synthesis and catabolism genes during root growth of Arabidopsis. Therefore, GCN5 and ADA2b are positive regulators of cytokinin signaling during root growth by modulating histone acetylation and the expression of genes involved in cytokinin synthesis and catabolism. Auxin application in the roots of wild-type seedlings increased TCS::GFP expression. In contrast, ada2b and ada2ada2b mutant plants do not show the auxin-induced TCS signal, suggesting that GCN5 and ADA2b are required for the auxin-induced cytokinin signaling in early root growth.