Merck
  • TNF antagonist sensitizes synovial fibroblasts to ferroptotic cell death in collagen-induced arthritis mouse models.
TNF antagonist sensitizes synovial fibroblasts to ferroptotic cell death in collagen-induced arthritis mouse models.
Nature communications (2022-02-05)
Jiao Wu, Zhuan Feng, Liang Chen, Yong Li, Huijie Bian, Jiejie Geng, Zhao-Hui Zheng, Xianghui Fu, Zhuo Pei, Yifei Qin, Liu Yang, Yilin Zhao, Ke Wang, Ruo Chen, Qian He, Gang Nan, Xuejun Jiang, Zhi-Nan Chen, Ping Zhu
ABSTRACT
Ferroptosis is a nonapoptotic cell death process that requires cellular iron and the accumulation of lipid peroxides. In progressive rheumatoid arthritis (RA), synovial fibroblasts proliferate abnormally in the presence of reactive oxygen species (ROS) and elevated lipid oxidation. Here we show, using a collagen-induced arthritis (CIA) mouse model, that imidazole ketone erastin (IKE), a ferroptosis inducer, decreases fibroblast numbers in the synovium. Data from single-cell RNA sequencing further identify two groups of fibroblasts that have distinct susceptibility to IKE-induced ferroptosis, with the ferroptosis-resistant fibroblasts associated with an increased TNF-related transcriptome. Mechanistically, TNF signaling promotes cystine uptake and biosynthesis of glutathione (GSH) to protect fibroblasts from ferroptosis. Lastly, low dose IKE together with etanercept, a TNF antagonist, induce ferroptosis in fibroblasts and attenuate arthritis progression in the CIA model. Our results thus imply that the combination of TNF inhibitors and ferroptosis inducers may serve as a potential candidate for RA therapy.
Sigma-Aldrich
Milli-Mark® Anti-Collagen Type I-FITC Antibody, clone 5D8-G9, clone 5D8-G9, Milli-Mark®, from mouse
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks