Merck
  • Paradoxical neuronal hyperexcitability in a mouse model of mitochondrial pyruvate import deficiency.
Paradoxical neuronal hyperexcitability in a mouse model of mitochondrial pyruvate import deficiency.
eLife (2022-02-22)
Andres De La Rossa, Marine H Laporte, Simone Astori, Thomas Marissal, Sylvie Montessuit, Preethi Sheshadri, Eva Ramos-Fernández, Pablo Mendez, Abbas Khani, Charles Quairiaux, Eric B Taylor, Jared Rutter, José Manuel Nunes, Alan Carleton, Michael R Duchen, Carmen Sandi, Jean-Claude Martinou
ABSTRACT
Neuronal excitation imposes a high demand of ATP in neurons. Most of the ATP derives primarily from pyruvate-mediated oxidative phosphorylation, a process that relies on import of pyruvate into mitochondria occuring exclusively via the mitochondrial pyruvate carrier (MPC). To investigate whether deficient oxidative phosphorylation impacts neuron excitability, we generated a mouse strain carrying a conditional deletion of MPC1, an essential subunit of the MPC, specifically in adult glutamatergic neurons. We found that, despite decreased levels of oxidative phosphorylation and decreased mitochondrial membrane potential in these excitatory neurons, mice were normal at rest. Surprisingly, in response to mild inhibition of GABA mediated synaptic activity, they rapidly developed severe seizures and died, whereas under similar conditions the behavior of control mice remained unchanged. We report that neurons with a deficient MPC were intrinsically hyperexcitable as a consequence of impaired calcium homeostasis, which reduced M-type potassium channel activity. Provision of ketone bodies restored energy status, calcium homeostasis and M-channel activity and attenuated seizures in animals fed a ketogenic diet. Our results provide an explanation for the seizures that frequently accompany a large number of neuropathologies, including cerebral ischemia and diverse mitochondriopathies, in which neurons experience an energy deficit.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-mtHsp70 Antibody, clone JG1, clone JG1, from mouse
Sigma-Aldrich
Kainic acid monohydrate, ≥99% (TLC)
Sigma-Aldrich
Anti-MPC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Mitochondrial Pyruvate Carrier 2 (BRP44), clone n/a, from mouse