Merck
  • Targeted dual inhibition of c-Met/VEGFR2 signalling by foretinib improves antitumour effects of nanoparticle paclitaxel in gastric cancer models.
Targeted dual inhibition of c-Met/VEGFR2 signalling by foretinib improves antitumour effects of nanoparticle paclitaxel in gastric cancer models.
Journal of cellular and molecular medicine (2021-05-04)
Meghan Grojean, Margaret A Schwarz, Johann R Schwarz, Sazzad Hassan, Urs von Holzen, Changhua Zhang, Roderich E Schwarz, Niranjan Awasthi
ABSTRACT
Elevated expression of multiple growth factors and receptors including c-Met and VEGFR has been reported in gastric adenocarcinoma (GAC) and thus provides a potentially useful therapeutic target. The therapeutic efficacy of foretinib, a c-Met/VEGFR2 inhibitor, was determined in combination with nanoparticle paclitaxel (NPT) in GAC. Animal studies were conducted in NOD/SCID mice in subcutaneous and peritoneal dissemination xenografts. The mechanism of action was assessed by Immunohistochemical and Immunoblot analyses. In c-Met overexpressing MKN-45 cell-derived xenografts, NPT and foretinib demonstrated inhibition in tumour growth, while NPT plus foretinib showed additive effects. In c-Met low-expressing SNU-1 or patient-derived xenografts, the foretinib effect was smaller, while NPT had a similar effect compared with MKN-45, as NPT plus foretinib still exhibited an additive response. Median mice survival was markedly improved by NPT (83%), foretinib (100%) and NPT plus foretinib (230%) in peritoneal dissemination xenografts. Subcutaneous tumour analyses exhibited that foretinib increased cancer cell death and decreased cancer cell proliferation and tumour vasculature. NPT and foretinib suppressed the proliferation of GAC cells in vitro and had additive effects in combination. Further, foretinib caused a dramatic decrease in phosphorylated forms of c-Met, ERK, AKT and p38. Foretinib led to a decrease in Bcl-2, and an increase in p27, Bax, Bim, cleaved PARP-1 and cleaved caspase-3. Thus, these findings highlight the antitumour impact of simultaneous suppression of c-Met and VEGFR2 signalling in GAC and its potential to enhance nanoparticle paclitaxel response. This therapeutic approach might lead to a clinically beneficial combination to increase GAC patients' survival.
Sigma-Aldrich
Anti-Endomucin Antibody, clone V.5C7, clone V.5C7, from rat