Merck
  • Carboxypeptidase A3 expression in canine mast cell tumors and tissue-resident mast cells.
Carboxypeptidase A3 expression in canine mast cell tumors and tissue-resident mast cells.
Veterinary pathology (2021-12-14)
Sanna Hämäläinen, Lauri Kareinen, Antti Sukura, Ilona Kareinen
ABSTRACT
Mast cell tumors (MCTs) are one of the most common cutaneous malignancies in dogs. Previous studies have reported expression of mast cell-specific proteases chymase and tryptase in canine cutaneous MCTs and in connective tissue and mucosal mast cells. In humans and rodents, mast cells express an additional specific protease, carboxypeptidase A3 (CPA3). In this article, we describe CPA3 immunoreactivity in connective tissue, visceral, mucosal, and neoplastic mast cells in dogs. Positive immunolabeling for CPA3 was observed in nonneoplastic mast cells in 20/20 formalin-fixed paraffin-embedded normal tissues (skin, liver, spleen, intestine), and in 63/63 MCTs irrespective of their histological grade. CPA3 protein expression was comparable to that of c-kit in both the nonneoplastic and neoplastic mast cells. Three distinct labeling patterns (membranous, diffuse, and focal cytoplasmic) were observed for CPA3 in MCTs. The focal cytoplasmic labeling pattern was associated with high-grade MCTs staged with the Kiupel 2-tier grading criteria. We propose CPA3 as a novel immunohistochemical marker for canine mast cells in health and disease.
Sigma-Aldrich
Anti-CPA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution