Merck
  • High glucose removes natural anti-α-galactoside and anti-β-glucoside antibody immune complexes adhering to surface O-glycoproteins of normal platelets and enhances platelet aggregation.
High glucose removes natural anti-α-galactoside and anti-β-glucoside antibody immune complexes adhering to surface O-glycoproteins of normal platelets and enhances platelet aggregation.
Experimental and therapeutic medicine (2021-12-23)
Sreedevi Karthi, Sangeetha Sukumari-Ramesh, Mandagini Geetha, Padinjaradath Sankunni Appukuttan
ABSTRACT
Human natural anti-α-galactoside (anti-Gal) and anti-β-glucoside (ABG) antibodies were previously reported to recognize the serine- and threonine-rich peptide sequences (STPS) of albumin-associated O-glycoproteins (AOP1 and AOP2) as surrogate antigens, forming anti-Gal/ABG-AOP1/AOP2-albumin triplet immune complexes in plasma. Since antibodies in these triplets still possessed unoccupied binding sites, the presence of triplets on human platelets that abound in surface O-glycoproteins was examined. Upon treatment with α-galactosides and β-glucosides, normal platelets freshly isolated from young healthy individuals released triplets identical with plasma triplets according to ELISA results. The resulting denuded platelets, unless pre-treated with fibrinogen or the O-glycan-binding lectin jacalin, recaptured these sugar-extracted triplets in the absence of antibody-specific sugars, suggesting that the triplet antibodies recognized the STPS of O-glycosylated receptors on platelets. Molecular weight of the dominant jacalin-binding subunit on triplet-free platelet membrane was 116 kDa, close to the ~120 kDa reported for the IIb subunit of the most abundant fibrinogen-binding platelet O-glycoprotein, GPIIb/IIIa. Denuded, but not native, platelets underwent slow spontaneous aggregation and rapid ADP-mediated GPIIb/IIIa-dependent aggregation according to spectrophotometric assay. Pre-treatment of denuded platelets with jacalin significantly reduced their ADP-mediated aggregation. Amyloid β (Aβ-42 monomer) was reported to bind triplet O-glycoproteins through their STPS but not to albumin or the antibodies. This peptide bound to the triplets on normal platelets and to surface membrane O-glycoproteins on denuded platelets, suggesting that the surface O-glycoproteins on the normal platelets were engaged and masked by the triplets. The ABG-specific sugar glucose denuded the platelets at concentrations typically reached in diabetic sera, since anti-Gal specific or ABG-specific sugar released the triplets of both the antibodies from the platelets. In conclusion, the present study offered rationale for the presence of anti-Gal/ABG-O-glycoprotein-albumin triplets on normal platelets, for the role of triplets in platelet physiology amidst circulating platelet-activating factors such as ADP, and for platelet vulnerability during diabetes.
Sigma-Aldrich
Anti-Human Albumin antibody produced in rabbit, whole antiserum