Merck
  • Nesprin-2 is a multi-isomeric protein that binds lamin and emerin at the nuclear envelope and forms a subcellular network in skeletal muscle.
Nesprin-2 is a multi-isomeric protein that binds lamin and emerin at the nuclear envelope and forms a subcellular network in skeletal muscle.
Journal of cell science (2005-01-27)
Qiuping Zhang, Cassandra D Ragnauth, Jeremy N Skepper, Nathalie F Worth, Derek T Warren, Roland G Roberts, Peter L Weissberg, Juliet A Ellis, Catherine M Shanahan
ABSTRACT
Nesprin-2 is a multi-isomeric, modular protein composed of variable numbers of spectrin-repeats linked to a C-terminal transmembrane domain and/or to N-terminal paired calponin homology (CH) domains. The smaller isoforms of nesprin-2 co-localize with and bind lamin A and emerin at the inner nuclear envelope (NE). In SW-13 cells, which lack lamin A/C, nesprin-2 epitopes and emerin were both mislocalized and formed aggregates in the endoplasmic reticulum (ER). The larger isoforms and other CH-domain-containing isoforms co-localize with heterochromatin within the nucleus and are also present at the outer NE and in multiple cytoplasmic compartments. Nesprin-2 isoforms relocalize during in vitro muscle differentiation of C2C12 myoblasts to the sarcomere of myotubes. Immunogold electron microscopy using antibodies specific for three different epitopes detected nesprin-2 isoforms at multiple locations including intranuclear foci, both membranes of the NE, mitochondria, sarcomeric structures and plasma membrane foci. In adult skeletal muscle, confocal immunolocalization studies demonstrated that nesprin-2 epitopes were present at the Z-line and were also associated with the sarcoplasmic reticulum (SR) in close apposition to SERCA2. These data suggest that nesprin-2 isoforms form a linking network between organelles and the actin cytoskeleton and thus may be important for maintaining sub-cellular spatial organisation. Moreover, its association at the NE with lamin and emerin, the genes mutated in Emery-Dreifuss muscular dystrophy, suggests a mechanism to explain how disruption of the NE leads to muscle dysfunction.
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Pricing and availability is not currently available.