Merck
  • Serum aflatoxin B1 antibody titer, percent hemolysis and transaminases in oral submucous fibrosis.
Serum aflatoxin B1 antibody titer, percent hemolysis and transaminases in oral submucous fibrosis.
Journal of oral and maxillofacial pathology : JOMFP (2021-08-06)
Rahul R Bhowate, Vidya K Lohe, Mrunal Ghusaji Meshram, Suwarna B Dangore
ABSTRACT
Areca nut is deeply rooted sociocultural habit in India. Areca nut reported to be infested by fungi during the field and storage conditions. Areca nut alkaloids, nitrosamines, tobacco and aflatoxin are cytotoxic, immunotoxic to red blood cell and epithelial cell. Hence, the present study was conducted to assess the serum aflatoxin B1 (AFB1) antibody titer, percent hemolysis and transaminases in oral submucous fibrosis (OSMF) patients. In this study, 128 participants of which 88 were suffering from OSMF. Twenty participants were areca nut habitual without OSMF (habitual control) and 20 participants without any habit (healthy control). For the detection of AFB1 antibody titer, AFB1 antigen (Sigma) A6636 from Aspergillus flavus was used. Percent hemolysis was estimated as per the procedure described by Mathuria and Verma. Serum Glutamic oxalo acetic transaminase (SGOT) and Serum Glutamic pyruvic transaminase (SGPT) were estimated by the optimized ultraviolet method using the enzyme-linked immunosorbent assay kit. Mean SGOT, SGPT, percent hemolysis and AFB1 antibody titer were significantly higher in participants with OSMF than the habitual and healthy controls. AFB1 antibody titer and % hemolysis in both OSMF and habitual without OSMF were showed significant correlation, i.e., increased AFB1 antibody titer with increased % hemolysis. A study result demonstrates that aflatoxin causes increase in serum transaminases which is indicative of liver damage in OSMF. The combined toxic effects of areca nut alkaloids, tobacco and AFB1 on red blood cell (RBC) cell wall might be responsible for increased percent hemolysis in OSMF and habitual control.
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Pricing and availability is not currently available.