Merck
  • Mild streptozotocin diabetes in the Göttingen minipig. A novel model of moderate insulin deficiency and diabetes.
Mild streptozotocin diabetes in the Göttingen minipig. A novel model of moderate insulin deficiency and diabetes.
American journal of physiology. Endocrinology and metabolism (2002-05-15)
Marianne O Larsen, Michael Wilken, Carsten F Gotfredsen, Richard D Carr, Ove Svendsen, Bidda Rolin
ABSTRACT
Nonrodent models of diabetes are needed for practical and physiological reasons. Induction of mild insulin-deficient diabetes was investigated in male Göttingen minipigs by use of streptozotocin (STZ) alone (75, 100, and 125 mg/kg) or 125 mg/kg combined with pretreatment with nicotinamide (NIA; 0, 20, 67, 100, 150, and 230 mg/kg). Use of NIA resulted in a less steep slope of the regression line between fasting plasma glucose and changing doses compared with STZ [-7.0 +/- 1.4 vs. 29.7 +/- 7.0 mM. mg(-1). kg(-1), P < 0.0001]. Intermediate NIA doses induced moderate changes of glucose tolerance [glucose area under the curve increased from 940 +/- 175 to 1,598 +/- 462 mM. min, P < 0.001 (100 mg/kg) and from 890 +/- 109 to 1,669 +/- 691 mM. min, P = 0.003 (67 mg/kg)] with reduced insulin secretion [1,248 +/- 602 pM. min after 16 days and 1,566 +/- 190 pM. min after 60 days vs. 3,251 +/- 804 pM. min in normal animals (P < 0.001)] and beta-cell mass [5.5 +/- 1.4 mg/kg after 27 days and 7.9 +/- 4.1 mg/kg after 60 days vs. 17.7 +/- 4.7 mg/kg in normal animals (P = 0.009)]. The combination of NIA and STZ provided a model characterized by fasting and especially postprandial hyperglycemia and reduced, but maintained, insulin secretion and beta-cell mass. This model holds promise as an important tool for studying the pathophysiology of diabetes and development of new pharmacological agents for treatment of the disease.
Sigma-Aldrich
Donkey serum
Pricing and availability is not currently available.