Merck
  • A protocol for generating germ-free Heligmosomoides polygyrus bakeri larvae for gnotobiotic helminth infection studies.
A protocol for generating germ-free Heligmosomoides polygyrus bakeri larvae for gnotobiotic helminth infection studies.
STAR protocols (2021-11-27)
Gabriel A Russell, Garrie Peng, Cynthia Faubert, Elena F Verdu, Siegfried Hapfelmeier, Irah L King
ABSTRACT
The microbes indigenous to helminth species are a major obstacle to deciphering host-parasite interactions. Repurposing a system of reversible bacterial colonization, we have generated germ-free Heligomosomoides polygyrus bakeri (Hpb) larvae that maintain the sterility of axenic mice upon infection. This protocol provides a valuable tool for controlled studies of helminth-microbiota-immune interactions.
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥85% (Vancomycin B)
Pricing and availability is not currently available.