Merck
  • Interstitial IgG antibody pharmacokinetics assessed by combined in vivo- and physiologically-based pharmacokinetic modelling approaches.
Interstitial IgG antibody pharmacokinetics assessed by combined in vivo- and physiologically-based pharmacokinetic modelling approaches.
The Journal of physiology (2017-09-30)
Miro J Eigenmann, Tine V Karlsen, Ben-Fillippo Krippendorff, Olav Tenstad, Ludivine Fronton, Michael B Otteneder, Helge Wiig
ABSTRACT
For therapeutic antibodies, total tissue concentrations are frequently reported as a lump sum measure of the antibody in residual plasma, interstitial fluid and cells. In terms of correlating antibody exposure to a therapeutic effect, however, interstitial pharmacokinetics might be more relevant. In the present study, we collected total tissue and interstitial antibody biodistribution data in mice and assessed the composition of tissue samples aiming to correct total tissue measurements for plasma and cellular content. All data and parameters were integrated into a refined physiologically-based pharmacokinetic model for monoclonal antibodies to enable the tissue-specific description of antibody pharmacokinetics in the interstitial space. We found that antibody interstitial concentrations are highly tissue-specific and dependent on the underlying capillary structure but, in several tissues, they reach relatively high interstitial concentrations, contradicting the still-prevailing view that both the distribution to tissues and the interstitial concentrations for antibodies are generally low. For most therapeutic antibodies, the interstitium is the target space. Although experimental methods for measuring antibody pharmacokinetics (PK) in this space are not well established, thus making quantitative assessment difficult, the interstitial antibody concentration is assumed to be low. In the present study, we combined direct quantification of antibodies in the interstitial fluid with a physiologically-based PK (PBPK) modelling approach, with the aim of better describing the PK of monoclonal antibodies in the interstitial space of different tissues. We isolated interstitial fluid by tissue centrifugation and conducted an antibody biodistribution study in mice, measuring total tissue and interstitial concentrations in selected tissues. Residual plasma, interstitial volumes and lymph flows, which are important PBPK model parameters, were assessed in vivo. We could thereby refine the PBPK modelling of monoclonal antibodies, better interpret antibody biodistribution data and more accurately predict their PK in the different tissue spaces. Our results indicate that, in tissues with discontinuous capillaries (liver and spleen), interstitial concentrations are reflected by the plasma concentration. In tissues with continuous capillaries (e.g. skin and muscle), ∼50-60% of the plasma concentration is found in the interstitial space. In the brain and kidney, on the other hand, antibodies are restricted to the vascular space. Our data may significantly impact the interpretation of biodistribution data of monoclonal antibodies and might be important when relating measured concentrations to a therapeutic effect. By contrast to the view that the antibody distribution to the interstitial space is limited, using direct measurements and model-based data interpretation, we show that high antibody interstitial concentrations are reached in most tissues.
Sigma-Aldrich
1,3,4,6-Tetrachloro-3α,6α-diphenylglycouril
Pricing and availability is not currently available.