Merck
  • CD4+ and CD8+ Circulating Memory T Cells Are Crucial in the Protection Induced by Vaccination with Salmonella Typhi Porins.
CD4+ and CD8+ Circulating Memory T Cells Are Crucial in the Protection Induced by Vaccination with Salmonella Typhi Porins.
Microorganisms (2021-05-01)
Luis Ontiveros-Padilla, Alberto García-Lozano, Araceli Tepale-Segura, Tania Rivera-Hernández, Rodolfo Pastelin-Palacios, Armando Isibasi, Lourdes A Arriaga-Pizano, Laura C Bonifaz, Constantino López-Macías
ABSTRACT
Salmonella enterica serovar Typhi (S. Typhi) porins, OmpC and OmpF, are potent inducers of the immune response against S. Typhi in mice and humans. Vaccination with porins induces the protection against 500 LD50 of S. Typhi, life-lasting bactericidal antibodies and effector T cell responses in mice; however, the nature of the memory T cell compartment and its contribution to protection remains unknown. In this work, we firstly observed that vaccination with porins induces in situ (skin) CD4+ and CD8+ T cell responses. Analysis of the porin-specific functional responses of skin CD4+ and CD8+ T cells showed IFN-gamma- and IL-17-producing cells in both T cell populations. The memory phenotype of porin-specific T cells indicated the presence of resident and effector memory phenotypes in the skin, and a central memory phenotype in the skin-draining lymph node. In addition, we demonstrated that vaccination with porins via skin reduces the bacterial burden following challenge. Finally, evaluating the role of the circulating T cell memory population in protection, we showed that circulating memory CD4+ and CD8+ T cells are crucial in porin-mediated protection against S. Typhi. Overall, this study highlights the importance of inducing circulating memory T cell responses in order to achieve the optimal protection provided by porins, showing a mechanism that could be sought in the rational development of vaccines.
Sigma-Aldrich
LB Broth with agar (Lennox), Highly-referenced microbial growth powder medium with Agar, low salt, suitable for salt-sensitive E. coli culture.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
LB Broth (Luria low salt), Powder microbial growth medium