Merck
  • The immune landscape of SARS-CoV-2-associated Multisystem Inflammatory Syndrome in Children (MIS-C) from acute disease to recovery.
The immune landscape of SARS-CoV-2-associated Multisystem Inflammatory Syndrome in Children (MIS-C) from acute disease to recovery.
iScience (2021-10-12)
Eleni Syrimi, Eanna Fennell, Alex Richter, Pavle Vrljicak, Richard Stark, Sascha Ott, Paul G Murray, Eslam Al-Abadi, Ashish Chikermane, Pamela Dawson, Scott Hackett, Deepthi Jyothish, Hari Krishnan Kanthimathinathan, Sean Monaghan, Prasad Nagakumar, Barnaby R Scholefield, Steven Welch, Naeem Khan, Sian Faustini, Kate Davies, Wioleta M Zelek, Pamela Kearns, Graham S Taylor
ABSTRACT
Multisystem inflammatory syndrome in children (MIS-C) is a life-threatening disease occurring several weeks after severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection. Deep immune profiling showed acute MIS-C patients had highly activated neutrophils, classical monocytes and memory CD8+ T-cells, with increased frequencies of B-cell plasmablasts and double-negative B-cells. Post treatment samples from the same patients, taken during symptom resolution, identified recovery-associated immune features including increased monocyte CD163 levels, emergence of a new population of immature neutrophils and, in some patients, transiently increased plasma arginase. Plasma profiling identified multiple features shared by MIS-C, Kawasaki Disease and COVID-19 and that therapeutic inhibition of IL-6 may be preferable to IL-1 or TNF-α. We identified several potential mechanisms of action for IVIG, the most commonly used drug to treat MIS-C. Finally, we showed systemic complement activation with high plasma C5b-9 levels is common in MIS-C suggesting complement inhibitors could be used to treat the disease.
Sigma-Aldrich
SIGMAFAST OPD, tablet