Merck
  • The release of toxic oligomers from α-synuclein fibrils induces dysfunction in neuronal cells.
The release of toxic oligomers from α-synuclein fibrils induces dysfunction in neuronal cells.
Nature communications (2021-03-24)
Roberta Cascella, Serene W Chen, Alessandra Bigi, José D Camino, Catherine K Xu, Christopher M Dobson, Fabrizio Chiti, Nunilo Cremades, Cristina Cecchi
ABSTRACT
The self-assembly of α-synuclein (αS) into intraneuronal inclusion bodies is a key characteristic of Parkinson's disease. To define the nature of the species giving rise to neuronal damage, we have investigated the mechanism of action of the main αS populations that have been observed to form progressively during fibril growth. The αS fibrils release soluble prefibrillar oligomeric species with cross-β structure and solvent-exposed hydrophobic clusters. αS prefibrillar oligomers are efficient in crossing and permeabilize neuronal membranes, causing cellular insults. Short fibrils are more neurotoxic than long fibrils due to the higher proportion of fibrillar ends, resulting in a rapid release of oligomers. The kinetics of released αS oligomers match the observed kinetics of toxicity in cellular systems. In addition to previous evidence that αS fibrils can spread in different brain areas, our in vitro results reveal that αS fibrils can also release oligomeric species responsible for an immediate dysfunction of the neurons in the vicinity of these species.
Sigma-Aldrich
Anti-alpha-Synuclein Antibody, oligomer-specific Syn33, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Amyloid Fibrils OC Antibody, serum, Chemicon®
Pricing and availability is not currently available.