Merck
  • Preterm birth prediction in asymptomatic women at mid-gestation using a panel of novel protein biomarkers: the Prediction of PreTerm Labor (PPeTaL) study.
Preterm birth prediction in asymptomatic women at mid-gestation using a panel of novel protein biomarkers: the Prediction of PreTerm Labor (PPeTaL) study.
American journal of obstetrics & gynecology MFM (2020-12-22)
San Min Leow, Megan K W Di Quinzio, Zhen Long Ng, Claire Grant, Tal Amitay, Ying Wei, Moshe Hod, Penelope M Sheehan, Shaun P Brennecke, Nir Arbel, Harry M Georgiou
ABSTRACT
Accurate prediction of spontaneous preterm labor/preterm birth in asymptomatic women remains an elusive clinical challenge because of the multi-etiological nature of preterm birth. The aim of this study was to develop and validate an immunoassay-based, multi-biomarker test to predict spontaneous preterm birth. This was an observational cohort study of women delivering from December 2017 to February 2019 at 2 maternity hospitals in Melbourne, Australia. Cervicovaginal fluid samples were collected from asymptomatic women at gestational week 16+0-24+0, and biomarker concentrations were quantified by enzyme-linked immunosorbent assay. Women were assigned to a training cohort (n = 136) and a validation cohort (n = 150) based on chronological delivery dates. Seven candidate biomarkers representing key pathways in utero-cervical remodeling were discovered by high-throughput bioinformatic search, and their significance in both in vivo and in vitro studies was assessed. Using a combination of the biomarkers for the first 136 women allocated to the training cohort, we developed an algorithm to stratify term birth (n = 124) and spontaneous preterm birth (n = 12) samples with a sensitivity of 100% (95% confidence interval, 76-100%) and a specificity of 74% (95% confidence interval, 66-81%). The algorithm was further validated in a subsequent cohort of 150 women (n = 139 term birth and n = 11 preterm birth), achieving a sensitivity of 91% (95% confidence interval, 62-100%) and a specificity of 78% (95% confidence interval, 70-84%). We have identified a panel of biomarkers that yield clinically useful diagnostic values when combined in a multiplex algorithm. The early identification of asymptomatic women at risk for preterm birth would allow women to be triaged to specialist clinics for further assessment and appropriate preventive treatment.
Sigma-Aldrich
Glyceryl trioleate, ≥99%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Carprofen, ≥97% (HPLC)
Sigma-Aldrich
Anti-Myosin IIA, non muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution