Merck
  • Horizontal Transfer of a Synthetic Metabolic Pathway between Plant Species.
Horizontal Transfer of a Synthetic Metabolic Pathway between Plant Species.
Current biology : CB (2017-09-26)
Yinghong Lu, Sandra Stegemann, Shreya Agrawal, Daniel Karcher, Stephanie Ruf, Ralph Bock
ABSTRACT
Transgene expression from the plastid (chloroplast) genome provides unique advantages, including high levels of foreign protein accumulation, convenient transgene stacking in operons, and increased biosafety due to exclusion of plastids from pollen transmission [1, 2]. However, applications in biotechnology and synthetic biology are severely restricted by the very small number of plant species whose plastid genomes currently can be transformed [3, 4]. Here we report a simple method for the introduction of useful plastid transgenes into non-transformable species. The transgenes tested comprised a synthetic operon encoding three components of a biosynthetic pathway for producing the high-value ketocarotenoid astaxanthin in the plastids of the cigarette tobacco, Nicotiana tabacum. Transplastomic N. tabacum plants accumulated astaxanthin to up to 1% of the plants' dry weight. We then used grafting, a procedure recently shown to facilitate horizontal genome transfer between plants [5-7], to let the transgenic chloroplast genome move across the graft junction from N. tabacum plants into plants of the nicotine-free tree species Nicotiana glauca. Transplastomic N. glauca trees expressing the synthetic pathway were recovered at high frequency, thus providing a straightforward method for extension of the transplastomic technology to new species.
Sigma-Aldrich
Violaxanthin, ≥90.0% (HPLC)
Pricing and availability is not currently available.