Merck
  • Optical isomers of 2-(2-ethoxyphenoxymethyl)tetrahydro-1,4-oxazine (viloxazine) and related compounds.
Optical isomers of 2-(2-ethoxyphenoxymethyl)tetrahydro-1,4-oxazine (viloxazine) and related compounds.
Journal of medicinal chemistry (1976-08-01)
R Howe, T Leigh, B S Rao, A H Todd
PMID966254
ABSTRACT
The optical isomers of 2-(2-ethoxyphenoxymethyl)tetrahydro-1,4-oxazine (viloxazine) and 2-(3-methoxyphenoxymethyl)tetrahydro-1,4-oxazine have been prepared and absolute configurations have been assigned. In their action on the central nervous system the S isomers are at least ten times more potent than the R isomers. The intermediate 4-benzyl-2-(p-toluensulfonyloxymethyl)tetrahydro-1,4-oxazine has been resolved. Its isomers provide a convenient starting point for the preparation of optical isomers of viloxazine analogues of known configuration.
Sigma-Aldrich
Viloxazine hydrochloride, ≥95% (HPLC)