Merck
  • N-methyl-2-pyridone-5-carboxamide (2PY)-Major Metabolite of Nicotinamide: An Update on an Old Uremic Toxin.
N-methyl-2-pyridone-5-carboxamide (2PY)-Major Metabolite of Nicotinamide: An Update on an Old Uremic Toxin.
Toxins (2016-11-18)
Aurélie Lenglet, Sophie Liabeuf, Sandra Bodeau, Loïc Louvet, Aurélien Mary, Agnès Boullier, Anne Sophie Lemaire-Hurtel, Alexia Jonet, Pascal Sonnet, Said Kamel, Ziad A Massy
ABSTRACT
N-methyl-2-pyridone-5-carboxamide (2PY, a major metabolite of nicotinamide, NAM) was recently identified as a uremic toxin. Recent interventional trials using NAM to treat high levels of phosphorus in end-stage renal disease have highlighted new potential uremic toxicities of 2PY. In the context of uremia, the accumulation of 2PY could be harmful-perhaps by inhibiting poly (ADP-ribose) polymerase-1 activity. Here, we review recently published data on 2PY's metabolism and toxicological profile.
Sigma-Aldrich
N-Methyl-4-pyridone-5-carboxamide